Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylaniline, >99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020678 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0678 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0678 Supplier MilliporeSigma Supelco Supplier No. 442327
Add to Cart
Add to Cart

2,6-Dimethylaniline is a alpha2-adrenergic agonist metabolite of xylazine, which can find applications as a sedative agent in food producing animals. It is also used in industries as a synthetic intermediate in the production of several products like dyes, pesticides and synthetic resins.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 87-62-7
Vapor Pressure <0.01 mmHg (20°C)
Density 0.984 g/mL (at 25°C)
Flash Point 91°C
Physical Form Neat
Linear Formula (CH3)2C6H3 NH2
MDL Number MFCD00007747
Melting Point 10°C to 12°C
Boiling Point 214°C (739 mmHg)
Synonym 2,6-Xylidine; 2-Amino-1,3-dimethylbenzene; 2-Amino-m-xylene
Type Organic Pollutant Standard
Quantity 1000 mg
Formula Weight 121.18
Percent Purity >99% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.