Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylaniline Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. X002925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X002925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X002925G Supplier TCI America Supplier No. X002925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity laboratory reagent with a minimum purity of 98.0%.

Chemical Identifiers

CAS 21436-98-6
Molecular Formula C8H12ClN
Molecular Weight (g/mol) 157.641
MDL Number MFCD00060213
InChI Key QNWMFJDRMZWZNN-UHFFFAOYSA-N
Synonym 2,6-Xylidine Hydrochloride
PubChem CID 88899
IUPAC Name 2,6-dimethylaniline;hydrochloride
SMILES CC1=C(C(=CC=C1)C)N.Cl

Specifications

Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 157.64
Percent Purity ≥98.0% (HPLC,N)
Grade Not Available
Chemical Name or Material 2,6-Dimethylaniline Hydrochloride
EINECSNumber (3)-0129
RTECSNumber ZF0375000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.