Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylbenzyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D35015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35015G 5 g
D350125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35015G Supplier TCI America Supplier No. D35015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5402-60-8
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.637
MDL Number MFCD01310815
InChI Key HPVRFWQMBYLJRL-UHFFFAOYSA-N
PubChem CID 221154
IUPAC Name 2-(chloromethyl)-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)CCl

Specifications

Color White-Yellow
Boiling Point 100°C
Quantity 5 g
UN Number 3261
Synonym 2,6-dimethylbenzyl chloride, 2-chloromethyl-1,3-dimethylbenzene, 2,6-dimethylbenzylchloride, 2,6-dimethyl benzylchloride, 1-chloromethyl-2,6-dimethylbenzene, 2-chloromethyl-1,3-dimethyl-benzene, 2-chloromethyl-m-xylene, benzene, 2-chloromethyl-1,3-dimethyl, 2, 6-dimethylbenzyl chloride
Formula Weight 154.64
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,6-Dimethylbenzyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.