Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylcyclohexanone (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. D128025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D128025ML 25 mL
D1280250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D128025ML Supplier TCI America Supplier No. D128025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2816-57-1
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00001637
InChI Key AILVYPLQKCQNJC-UHFFFAOYSA-N
Synonym 2,6-dimethylcyclohexanone, cyclohexanone, 2,6-dimethyl, cyclohexanone,6-dimethyl, 2,6-dimethyl cyclohexanone, 2,6-dimethyl-cyclohexanone, cyclohexanone,2,6-dimethyl, 2,6-dimethylcyclohexanone,c&t, 2,6-dimethyl-cyclohexan-1-one, 2,6-dimethylcyclohexanone cis + trans, 2,6-dimethylcyclohexanone, cis + trans
PubChem CID 17780
IUPAC Name 2,6-dimethylcyclohexan-1-one
SMILES CC1CCCC(C1=O)C

Specifications

Color Yellow
Quantity 25 mL
UN Number 1224
Formula Weight 126.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Dimethylcyclohexanone (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.