Learn More
Medchemexpress LLC 2,6-Dimethylhydroquinone | 654-42-2 | 138.17 | 500 MG
SDP
Catalog No. 5000446349
Encompass_Preferred
2,6-Dimethylhydroquinone is a key metabolic intermediate for the Mycobacterium strain DM1 during the degradation of 2,6-dimethylphenol (2,6-xylenol). It can serve as an indicator for early failures in biological treatment systems.
  • Key metabolic intermediate
  • Involved in degradation of 2,6-dimethylphenol
  • Indicator for early failures in biological treatment systems
Catalog No. 50-004-46349 Supplier Medchemexpress LLC Supplier No. HYW004874500MG
2,6-Dimethylhydroquinone is a key metabolic intermediate for the Mycobacterium strain DM1 during the degradation of 2,6-dimethylphenol (2,6-xylenol). It can serve as an indicator for early failures in biological treatment systems.
  • Key metabolic intermediate
  • Involved in degradation of 2,6-dimethylphenol
  • Indicator for early failures in biological treatment systems