Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylnaphthalene, TCI America™
SDP

Catalog No. D07511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D07511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D07511G Supplier TCI America Supplier No. D07511G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 581-42-0
Molecular Formula C12H12
Molecular Weight (g/mol) 156.228
MDL Number MFCD00004120
InChI Key YGYNBBAUIYTWBF-UHFFFAOYSA-N
Synonym naphthalene, 2,6-dimethyl, dimethylnaphthalene, mixture of isomers, 2,6-dmn, 2,6-dimethyl-naphthalene, unii-76u29qw3fm, 2,6-dimethylphthalene, 2,6-dimethylnaphthalen, naphthalene, 2,6-or 2,7-dimethyl, 2.6-dimethylnaphthalene, acmc-1arta
PubChem CID 11387
ChEBI CHEBI:34251
IUPAC Name 2,6-dimethylnaphthalene
SMILES CC1=CC2=C(C=C1)C=C(C=C2)C

Specifications

Melting Point 111°C
Color White
Quantity 1 g
Formula Weight 156.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethylnaphthalene
EINECSNumber (4)-0080
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.