Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylphenol, 99%

Catalog No. AAA1194736
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1194736 500 g
AAA119470E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1194736 Supplier Thermo Scientific Chemicals Supplier No. A1194736
  • $59.70
    $50.65 / Each of 1

    Save $9.05

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 576-26-1 | C8H10O | 122.17 g/mol

2,6-dimethylphenol is used in the synthesis of anti-oxidant compounds due to the phenol moiety in the structure. In addition, this compound is used as a reactant in the synthesis of polyphenylene ether polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-dimethylphenol is used in the synthesis of anti-oxidant compounds due to the phenol moiety in the structure. In addition, this compound is used as a reactant in the synthesis of polyphenylene ether polymers.

Solubility
Soluble in water and chloroform.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from strong bases and oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 576-26-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002240
InChI Key NXXYKOUNUYWIHA-UHFFFAOYSA-N
Synonym 2,6-xylenol, phenol, 2,6-dimethyl, 2,6-dimethyl phenol, 2-hydroxy-m-xylene, 1-hydroxy-2,6-dimethylbenzene, vic-m-xylenol, 2,6-dmp, 2,6-dimethyl-phenol, xylenol 235, 2-hydroxy-1,3-dimethylbenzene
PubChem CID 11335
IUPAC Name 2,6-dimethylphenol
SMILES CC1=CC=CC(C)=C1O

Specifications

Melting Point 44°C to 48°C
Density 1.15
Boiling Point 203°C
Flash Point 78°C (172°F)
Quantity 500 g
UN Number UN2261
Beilstein 1446677
Sensitivity Light sensitive
Merck Index 14,10082
Solubility Information Soluble in water and chloroform.
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material 2,6-Dimethylphenol
Show More Show Less
Hazard Category H301+H311-H314
Hazard Statement GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID
EINECSNumber 209-400-1
RTECSNumber ZE6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.