Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[(2,6-Dimethylphenyl)amino](oxo)acetic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7128186 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42651G 1 g
D42655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42651G Supplier TCI America Supplier No. D42651G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2903-48-2
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD10018453
InChI Key JFTAPSBIODPQJN-UHFFFAOYSA-N
Synonym (2,6-Dimethylanilino)(oxo)acetic Acid, N-(2,6-Dimethylphenyl)oxamic Acid, DMPAO
PubChem CID 14987869
IUPAC Name 2-(2,6-dimethylanilino)-2-oxoacetic acid
SMILES CC1=C(C(=CC=C1)C)NC(=O)C(=O)O

Specifications

Melting Point 173°C
Color White
Quantity 1 g
Formula Weight 193.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material [(2,6-Dimethylphenyl)amino](oxo)acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.