Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,6-Dimethylphenylmagnesium Bromide Solution

Catalog No. 425508100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-550-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 42-550-8100ML Supplier Sigma Aldrich Supplier No. 425508100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,6-Dimethylphenylmagnesium bromide can be used:As an intermediate in the synthesis of dibutyl(5,10-dihydro-1,9-di-p-toluoyl-5-p-tolyldipyrrinato)tin(IV), which is further used in the preparation of zinc-based porphyrin.As a substrate in the preparation of substituted fluorenes by reacting with 2-substituted arenesulfonates and dihaloarenes.

Specifications

CAS 1643-19-2
Density 1.039 g/mL at 25°C
Boiling Point 102°C
Molecular Formula C16H36BrN
Linear Formula (CH3CH2CH2CH2)4N(Br)
MDL Number MFCD00011633
Quantity 100 mL
Molecular Weight (g/mol) 322.37 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.