Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylpiperazine, MP Biomedicals

Catalog No. ICN204352 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204352 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204352 Supplier MP Biomedicals, Inc Supplier No. 0520435201

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-49-6
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD07772435,MFCD00005956
InChI Key IFNWESYYDINUHV-UHFFFAOYNA-N
Synonym piperazine, 2,6-dimethyl, 3,5-dimethylpiperazine, 2,6-dimethyl-piperazine, 2,6-dimethyl piperazine, 2r,6s-rel-2,6-dimethylpiperazine, pubchem8571, 2,6 dimethylpiperazine, 2,6-dimehtylpiperazine, piperazine,6-dimethyl, acmc-1bolo
PubChem CID 66056
IUPAC Name 2,6-dimethylpiperazine
SMILES CC1CNCC(C)N1

Specifications

Quantity 1 g
Formula Weight 114.19
Chemical Name or Material 2, 6-Dimethylpiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.