Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylpyrazine, 99%

Catalog No. p-7027712
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2468714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2468714 Supplier Thermo Scientific Chemicals Supplier No. B2468714
Only null left
Add to Cart

CAS: 108-50-9 | C6H8N2 | 108.144 g/mol

2,6-Dimethylpyrazine is used Preparation of chocolate, coffee, meat and nuts and other flavors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylpyrazine is used Preparation of chocolate, coffee, meat and nuts and other flavors

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-50-9
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006148
InChI Key HJFZAYHYIWGLNL-UHFFFAOYSA-N
Synonym pyrazine, 2,6-dimethyl, 3,5-dimethylpyrazine, 2,6-dimethylpiazine, 2,6-dimethylparadiazine, 2,6-dimethyl-1,4-diazine, ccris 2930, unii-n77q72c9i3, fema no. 3273, natural, 2,6-dimethyl pyrazine
PubChem CID 7938
IUPAC Name 2,6-dimethylpyrazine
SMILES CC1=CN=CC(=N1)C

Specifications

Melting Point 38°C to 43°C
Boiling Point 153°C to 154°C
Flash Point 52°C (125°F)
Odor Aromatic
Refractive Index 1.5
Quantity 25 g
UN Number UN1325
Beilstein 1726
Solubility Information Soluble in water.
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2,6-Dimethylpyrazine
Show More Show Less
Hazard Category H228-H302
Hazard Statement GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 203-589-4
RTECSNumber UQ2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.