Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylpyrazine 98.0+%, TCI America™
SDP

Catalog No. D152725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D152725G 25 g
D15275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D152725G Supplier TCI America Supplier No. D152725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108-50-9
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006148
InChI Key HJFZAYHYIWGLNL-UHFFFAOYSA-N
Synonym pyrazine, 2,6-dimethyl, 3,5-dimethylpyrazine, 2,6-dimethylpiazine, 2,6-dimethylparadiazine, 2,6-dimethyl-1,4-diazine, ccris 2930, unii-n77q72c9i3, fema no. 3273, natural, 2,6-dimethyl pyrazine
PubChem CID 7938
IUPAC Name 2,6-dimethylpyrazine
SMILES CC1=CN=CC(=N1)C

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 155°C
Quantity 25 g
UN Number 1325
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2,6-Dimethylpyrazine
EINECSNumber (5)-0950
RTECSNumber UQ2975000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.