Learn More
Medchemexpress LLC 2,6-dimethylpyridin-1-ium-1-olate | 1073-23-0 | MFCD00039715 | >98.0% | 123.15 g/mol | C7H9NO | 5 G
SDP
Catalog No. 5000284349
Encompass_Preferred
2,6-Lutidine N-oxide is an organic reagent (C7H9NO, MW 123.15 g/mol) commonly used as a ligand, oxidizing agent, and intermediate in organic synthesis and laboratory research. It is typically supplied as a colorless to light yellow liquid with high purity suitable for bench-scale reactions and analytical work.
  • High purity suitable for research and synthesis.
  • Useful as a ligand and mild oxidizing reagent.
  • Compatible with common organic solvents.
  • Supplied in small laboratory quantities for convenient handling and storage.
  • Identified by CAS 1073-23-0 for unambiguous substance reference.
Catalog No. 50-002-84349 Supplier Medchemexpress LLC Supplier No. HYW5397995G
2,6-Lutidine N-oxide is an organic reagent (C7H9NO, MW 123.15 g/mol) commonly used as a ligand, oxidizing agent, and intermediate in organic synthesis and laboratory research. It is typically supplied as a colorless to light yellow liquid with high purity suitable for bench-scale reactions and analytical work.
  • High purity suitable for research and synthesis.
  • Useful as a ligand and mild oxidizing reagent.
  • Compatible with common organic solvents.
  • Supplied in small laboratory quantities for convenient handling and storage.
  • Identified by CAS 1073-23-0 for unambiguous substance reference.