Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2,6-dimethylpyridin-4-yl)-[4-[7-(4-hydroxycyclohexyl)-2-[[(2S)-pentan-2-yl]amino]pyrrolo[2,3-d]pyrimi | 2226789-82-6 | 99.9% | 518.69 | C30H42N6O2 | 5 MG
SDP

Catalog No. 5000262962
Encompass_Preferred

UNC5293 is a potent, orally available small-molecule inhibitor selective for the MER receptor tyrosine kinase (MERTK). It demonstrates subnanomolar biochemical potency, cellular activity in B-ALL models, and favorable mouse pharmacokinetic properties; intended for research use.

  • Biochemical Ki = 190 pM and MERTK IC50 = 0.9 nM.
  • Cellular MERTK phosphorylation IC50 = 9.4 nM in B-ALL cells.
  • Selective over Axl, Tyro3, and Flt3; less potent versus FLT3 (IC50 = 170 nM).
  • Molecular weight 518.69 and formula C30H42N6O2.
  • Purity 99.93% and supplied as a 5 mg pack.
  • Exhibits favorable mouse pharmacokinetic properties and utility in leukemia research.

Catalog No. 50-002-62962 Supplier Medchemexpress LLC Supplier No. HY1322005MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UNC5293 is a potent, orally available small-molecule inhibitor selective for the MER receptor tyrosine kinase (MERTK). It demonstrates subnanomolar biochemical potency, cellular activity in B-ALL models, and favorable mouse pharmacokinetic properties; intended for research use.

  • Biochemical Ki = 190 pM and MERTK IC50 = 0.9 nM.
  • Cellular MERTK phosphorylation IC50 = 9.4 nM in B-ALL cells.
  • Selective over Axl, Tyro3, and Flt3; less potent versus FLT3 (IC50 = 170 nM).
  • Molecular weight 518.69 and formula C30H42N6O2.
  • Purity 99.93% and supplied as a 5 mg pack.
  • Exhibits favorable mouse pharmacokinetic properties and utility in leukemia research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.