Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diphenylbenzo[1,2-b:4,5-b']difuran, TCI America™
SDP

Catalog No. D4633200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4633200MG 200 mg
D46331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4633200MG Supplier TCI America Supplier No. D4633200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5379-77-1
Molecular Formula C22H14O2
Molecular Weight (g/mol) 310.352
InChI Key QDHGBWBEBOMJCI-UHFFFAOYSA-N
PubChem CID 67295972
IUPAC Name 2,6-diphenylfuro[2,3-f][1]benzofuran
SMILES C1=CC=C(C=C1)C2=CC3=CC4=C(C=C(O4)C5=CC=CC=C5)C=C3O2

Specifications

Melting Point 345°C
Color White-Yellow
Quantity 200 mg
Formula Weight 310.35
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diphenylbenzo[1,2-b:4,5-b′]difuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.