Learn More
Medchemexpress LLC 2,6-dithiopurine | 5437-25-2 | 98.0% | 184.25 g/mol | C5H4N4S2 | 250 MG
SDP
Catalog No. 5000302573
Encompass_Preferred
2,6-Dithiopurine is a nucleophilic scavenger that reacts with electrophilic carcinogens to diminish DNA adduct formation and inhibit the initiation of carcinogenesis. It is intended for research use and is available as a solid and as DMSO solutions for experimental flexibility.
  • Acts as a nucleophilic scavenger against electrophilic carcinogens.
  • Reduces DNA adduct formation and carcinogenesis initiation.
  • Suitable for biochemical and carcinogenesis research applications.
  • Available in solid and solution formats for flexible use.
  • High purity (≥98.0%) supports reproducible experimental results.
Catalog No. 50-003-02573 Supplier Medchemexpress LLC Supplier No. 250MGHYW016286
2,6-Dithiopurine is a nucleophilic scavenger that reacts with electrophilic carcinogens to diminish DNA adduct formation and inhibit the initiation of carcinogenesis. It is intended for research use and is available as a solid and as DMSO solutions for experimental flexibility.
  • Acts as a nucleophilic scavenger against electrophilic carcinogens.
  • Reduces DNA adduct formation and carcinogenesis initiation.
  • Suitable for biochemical and carcinogenesis research applications.
  • Available in solid and solution formats for flexible use.
  • High purity (≥98.0%) supports reproducible experimental results.