Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI America 2,6-Lutidine, Reagent, 98% Purity, 25 mL

Catalog No. 502780984
Encompass_Preferred

2,6-Lutidine, also known as 2,6-dimethylpyridine, is a heterocyclic organic compound widely utilized in chemical research and industrial applications. This reagent is particularly noted for its use in protecting alcohols through tert-butyldiphenylsilyl (TBDPS) triflate chemistry.

  • Purity: >98.0% (GC)
  • Molecular formula: C7H9N
  • Molecular weight: 107.16 g/mol
  • Physical state: Clear liquid
  • Boiling point: 143°C to 145°C
  • Melting point: -6°C
  • Refractive index: 1.50
  • Storage: Room temperature, store under inert gas in a cool, dark place

Catalog No. 50-278-0984 Supplier Grainger Supplier No. 19KM65
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,6-Lutidine, also known as 2,6-dimethylpyridine, is a heterocyclic organic compound widely utilized in chemical research and industrial applications. This reagent is particularly noted for its use in protecting alcohols through tert-butyldiphenylsilyl (TBDPS) triflate chemistry.

  • Purity: >98.0% (GC)
  • Molecular formula: C7H9N
  • Molecular weight: 107.16 g/mol
  • Physical state: Clear liquid
  • Boiling point: 143°C to 145°C
  • Melting point: -6°C
  • Refractive index: 1.50
  • Storage: Room temperature, store under inert gas in a cool, dark place

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.