Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,6-Naphthalenedisulfonic acid disodium salt

Catalog No. N605100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N605100G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N605100G Supplier Sigma Aldrich Supplier No. N605100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl 4,4,4-trifluoro-2-butynoate is an unsymmetrical internal alkyne.ethyl 4,4,4-trifluoro-2-butynoate has been used to investigate the regioselectivity of the insertion reaction with cyclometalated iridium and rhodium complexes. it may be used in the synthesis of the following compounds :
  • (2s,4ar,6ar,7r,9s,10as,10br)-methyl 9-acetoxy-2-(5-(ethoxycarbonyl)-6-(trifluoromethyl)-7-oxabicyclo[2.2.1]hepta-2,5-dien-2-yl)-6a,10b-dimethyl-4,10-dioxododecahydro-1h-benzo[f]isochromene-7-carboxylate
  • (2s,4ar,6ar,7r,9s,10as,10br)-methyl 9-acetoxy-2-(6-(ethoxycarbonyl)-5-(trifluoromethyl)-7-oxabicyclo[2.2.1]hepta-2,5-dien-2-yl)-6a,10b-dimethyl-4,10-dioxododecahydro-1hbenzo[f]isochromene-7-carboxylate

Specifications

CAS 1655-45-4
Melting Point >300°C (lit.)
Molecular Formula C10H6Na2O6S2
Linear Formula C10H6(SO3Na)2
MDL Number MFCD00004092
Quantity 500 mg
Molecular Weight (g/mol) 332.26
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.