Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,6-Nonadienoic acid, 9-(3,3-dimethyl-2-oxiranyl)-3,7-dimethyl-, methyl-d3 ester, (2E,6E)- | 951116-89-5 | 2.5 MG
SDP

Catalog No. 5000418020
Encompass_Preferred

Juvenile Hormone III-d3 is the deuterium labeled Juvenile Hormone III. Stable heavy isotopes like deuterium are incorporated into drug molecules as tracers for quantitation during drug development. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs. This product is for research use only and has not been fully validated for medical applications.

  • Deuterium labeled Juvenile Hormone III
  • Used as tracer for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Molecular formula: C16H23D3O3
  • Molecular weight: 269.39
  • Appearance: Colorless to light yellow oil
  • Purity (GC): 97.85%
  • Isotopic enrichment: 99.6%

Catalog No. 50-004-18020 Supplier Medchemexpress LLC Supplier No. HYN7240S2.5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Juvenile Hormone III-d3 is the deuterium labeled Juvenile Hormone III. Stable heavy isotopes like deuterium are incorporated into drug molecules as tracers for quantitation during drug development. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs. This product is for research use only and has not been fully validated for medical applications.

  • Deuterium labeled Juvenile Hormone III
  • Used as tracer for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Molecular formula: C16H23D3O3
  • Molecular weight: 269.39
  • Appearance: Colorless to light yellow oil
  • Purity (GC): 97.85%
  • Isotopic enrichment: 99.6%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.