Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,6-Octadien-1-ol, 3,7-dimethyl-, 1-acetate, (2Z)- | 141-12-8 | MFCD00063205 | 99.0% | 196.29 | C12H20O2 | 500 MG
SDP

Catalog No. 5000302907
Encompass_Preferred

Neryl acetate is a monoterpene acetate ester isolated from citrus and other plant essential oils. It is used as an analytical standard, fragrance ingredient, and research reagent, offered as a high-purity liquid suitable for analytical, formulation, and laboratory applications.

  • High purity: 98.99% (GC).
  • Colorless to light yellow liquid with density ~0.91 g/cm3.
  • Molecular formula C12H20O2; molecular weight 196.29.
  • Available in small quantities and solution formats for analytical use.
  • Recommended storage: -20°C (pure) and -80°C (in solvent) for long-term stability.

Catalog No. 50-003-02907 Supplier Medchemexpress LLC Supplier No. HYW014983500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Neryl acetate is a monoterpene acetate ester isolated from citrus and other plant essential oils. It is used as an analytical standard, fragrance ingredient, and research reagent, offered as a high-purity liquid suitable for analytical, formulation, and laboratory applications.

  • High purity: 98.99% (GC).
  • Colorless to light yellow liquid with density ~0.91 g/cm3.
  • Molecular formula C12H20O2; molecular weight 196.29.
  • Available in small quantities and solution formats for analytical use.
  • Recommended storage: -20°C (pure) and -80°C (in solvent) for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.