Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,6-Octadien-1-ol, 3,7-dimethyl-, (2E)- | 106-24-1 | 250 MG
SDP

Catalog No. 5000420691
Encompass_Preferred

Geraniol is an olefin terpene with oral activity, known for inhibiting cell proliferation and promoting apoptosis. It exhibits a broad spectrum of biological activities, including antibacterial, antifungal, antioxidant, anti-inflammatory, and antitumor effects, making it a valuable compound for various research applications, particularly in diabetes studies.

  • Inhibits cell proliferation and promotes apoptosis
  • Possesses antibacterial and antifungal activities
  • Offers antioxidant and anti-inflammatory benefits
  • Demonstrates antitumor properties
  • Useful in diabetes research
  • Molecular weight: 154.25
  • Molecular formula: C10H18O
  • Purity: 99.79%
  • Appearance: Colorless to light yellow liquid
  • Recommended storage: 4°C, protected from light; in solvent: -80°C for 6 months, or -20°C for 1 month (protected from light)

Catalog No. 50-004-20691 Supplier Medchemexpress LLC Supplier No. 250MGHYN6952
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Geraniol is an olefin terpene with oral activity, known for inhibiting cell proliferation and promoting apoptosis. It exhibits a broad spectrum of biological activities, including antibacterial, antifungal, antioxidant, anti-inflammatory, and antitumor effects, making it a valuable compound for various research applications, particularly in diabetes studies.

  • Inhibits cell proliferation and promotes apoptosis
  • Possesses antibacterial and antifungal activities
  • Offers antioxidant and anti-inflammatory benefits
  • Demonstrates antitumor properties
  • Useful in diabetes research
  • Molecular weight: 154.25
  • Molecular formula: C10H18O
  • Purity: 99.79%
  • Appearance: Colorless to light yellow liquid
  • Recommended storage: 4°C, protected from light; in solvent: -80°C for 6 months, or -20°C for 1 month (protected from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.