Learn More
Medchemexpress LLC 2,6-Octadien-1-ol, 3,7-dimethyl-, (2E)- | 106-24-1 | MFCD00002917 | 1 ML
SDP
Catalog No. 5000418338
Encompass_Preferred
Geraniol is an olefin terpene that exhibits oral activity. It inhibits cell proliferation, promotes apoptosis, and demonstrates antibacterial, antifungal, antioxidant, anti-inflammatory, and antitumor properties. This compound can also be utilized for research related to diabetes.
  • Inhibits cell proliferation and promotes apoptosis
  • Exhibits antibacterial and antifungal activities
  • Demonstrates antioxidant and anti-inflammatory properties
  • Possesses antitumor activities
  • Useful in studying diabetes
  • Blocks the cell cycle and decreases cell growth
  • Increases G1 and/or sub-G1 phase cells
  • Increases caspase-3 activity
  • Inhibits tumor growth in PC-3 nude mouse models
  • Improves hyperglycemia
  • Alleviates oxidative stress in hepatopancreatic toxicity
Catalog No. 50-004-18338 Supplier Medchemexpress LLC Supplier No. HYN695210MM/1ML
Geraniol is an olefin terpene that exhibits oral activity. It inhibits cell proliferation, promotes apoptosis, and demonstrates antibacterial, antifungal, antioxidant, anti-inflammatory, and antitumor properties. This compound can also be utilized for research related to diabetes.
  • Inhibits cell proliferation and promotes apoptosis
  • Exhibits antibacterial and antifungal activities
  • Demonstrates antioxidant and anti-inflammatory properties
  • Possesses antitumor activities
  • Useful in studying diabetes
  • Blocks the cell cycle and decreases cell growth
  • Increases G1 and/or sub-G1 phase cells
  • Increases caspase-3 activity
  • Inhibits tumor growth in PC-3 nude mouse models
  • Improves hyperglycemia
  • Alleviates oxidative stress in hepatopancreatic toxicity