Learn More
Medchemexpress LLC 2,6-octadienal, 3,7-dimethyl-, (2Z)- | 106-26-3 | MFCD00213902 | 97.9% | 152.23 g/mol | C10H16O | 25mg
SDP
Catalog No. 5000268315
Encompass_Preferred
Neral is a monoterpene compound that has anti-inflammatory and anti-cancer properties Neral can inhibit TNF- and IL-6 and it also suppresses inflammatory mediators like pro-IL-1 iNOS COX-2 and NLRP-3[1][2]
Catalog No. 50-002-68315 Supplier Medchemexpress LLC Supplier No. HYW92318925MG
Neral is a monoterpene compound that has anti-inflammatory and anti-cancer properties Neral can inhibit TNF- and IL-6 and it also suppresses inflammatory mediators like pro-IL-1 iNOS COX-2 and NLRP-3[1][2]