Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Octadiyne 97.0+%, TCI America™
SDP

Catalog No. O019401ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O019401ML 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O019401ML Supplier TCI America Supplier No. O01940.1ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 764-73-8
Molecular Formula C8H10
Molecular Weight (g/mol) 106.168
MDL Number MFCD00041620
InChI Key PHNFSGFUJPBCLS-UHFFFAOYSA-N
PubChem CID 136585
IUPAC Name octa-2,6-diyne
SMILES CC#CCCC#CC

Specifications

Quantity 0.1 mL
Formula Weight 106.17
Percent Purity ≥97.0% (GC)
Physical Form Crystals
Chemical Name or Material 2,6-Octadiyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.