Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,6-piperidinedione, 3-(1,3-dihydro-5-iodo-1-oxo-2H-isoindol-2-yl) | 2291364-01-5 | MFCD32853043 | 98.0% | 370.14 | C13H11IN2O3 | 50 MG
SDP

Catalog No. 5000281714
Encompass_Preferred

Deoxy-thalidomide-5-iodine is an iodinated thalidomide analog intended for chemical biology and PROTAC applications; it functions as a cereblon-binding ligand for E3 ligase recruitment and is supplied with analytical data confirming identity and high purity.

  • Iodinated thalidomide analog useful for PROTAC research.
  • Binds cereblon to enable E3 ligase recruitment.
  • High purity: 98.0% by NMR.
  • Molecular weight 370.14; formula C13H11IN2O3.
  • Appearance: blue to blue-green solid.
  • Store protected from light at 4°C; in solvent keep at -80°C (6 months) or -20°C (1 month).
  • Supplied in small research quantities (50 mg).

Catalog No. 50-002-81714 Supplier Medchemexpress LLC Supplier No. HYW44127150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deoxy-thalidomide-5-iodine is an iodinated thalidomide analog intended for chemical biology and PROTAC applications; it functions as a cereblon-binding ligand for E3 ligase recruitment and is supplied with analytical data confirming identity and high purity.

  • Iodinated thalidomide analog useful for PROTAC research.
  • Binds cereblon to enable E3 ligase recruitment.
  • High purity: 98.0% by NMR.
  • Molecular weight 370.14; formula C13H11IN2O3.
  • Appearance: blue to blue-green solid.
  • Store protected from light at 4°C; in solvent keep at -80°C (6 months) or -20°C (1 month).
  • Supplied in small research quantities (50 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.