Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,6-piperidinedione, 3-(1,3-dihydro-5-iodo-1-oxo-2H-isoindol-2-yl) | 2291364-01-5 | MFCD32853043 | 98.0% | 370.14 g·mol⁻¹ | C13H11IN2O3 | 1 G
SDP

Catalog No. 5000281712
Encompass_Preferred

Deoxy-thalidomide-5-iodine is a thalidomide analogue used as an E3 ligase ligand in PROTAC research and cereblon-targeting studies. Supplied as a high-purity solid, it is commonly used as a building block for synthesis of degraders and chemical probes.

  • High purity suitable for research (98.0%).
  • Chemical formula C13H11IN2O3.
  • Molecular weight 370.14 g·mol⁻¹.
  • Useful as an E3 ligase (cereblon) ligand in PROTAC design.
  • Solid form for ease of handling and storage.
  • Store protected from light at refrigerated temperatures; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-81712 Supplier Medchemexpress LLC Supplier No. HYW4412711G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deoxy-thalidomide-5-iodine is a thalidomide analogue used as an E3 ligase ligand in PROTAC research and cereblon-targeting studies. Supplied as a high-purity solid, it is commonly used as a building block for synthesis of degraders and chemical probes.

  • High purity suitable for research (98.0%).
  • Chemical formula C13H11IN2O3.
  • Molecular weight 370.14 g·mol⁻¹.
  • Useful as an E3 ligase (cereblon) ligand in PROTAC design.
  • Solid form for ease of handling and storage.
  • Store protected from light at refrigerated temperatures; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.