Learn More
Medchemexpress LLC 2,6-piperidinedione, 3-(1,3-dihydro-5-iodo-1-oxo-2h-isoindol-2-yl)- | 2291364-01-5 | MFCD32853043 | 98.0% | 370.14 g/mol | C13H11IN2O3 | 25 MG
SDP
Catalog No. 5000281713
Encompass_Preferred
Deoxy-thalidomide-5-iodine is a thalidomide analog bearing an iodine substituent at the 5-position used as a cereblon-binding ligand in PROTAC and targeted protein degradation research. Supplied as a colored solid with high reported purity, it is intended for chemical biology and linker-conjugation workflows.
  • Thalidomide-derived cereblon ligand suitable for PROTAC design.
  • High reported purity (98.0% by NMR) for research applications.
  • Blue to blue-green solid physical form for easy handling.
  • Molecular formula C13H11IN2O3 and molecular weight 370.14 g/mol.
  • Suitable for linker attachment and synthesis of bifunctional degraders.
Catalog No. 50-002-81713 Supplier Medchemexpress LLC Supplier No. HYW44127125MG
Deoxy-thalidomide-5-iodine is a thalidomide analog bearing an iodine substituent at the 5-position used as a cereblon-binding ligand in PROTAC and targeted protein degradation research. Supplied as a colored solid with high reported purity, it is intended for chemical biology and linker-conjugation workflows.
  • Thalidomide-derived cereblon ligand suitable for PROTAC design.
  • High reported purity (98.0% by NMR) for research applications.
  • Blue to blue-green solid physical form for easy handling.
  • Molecular formula C13H11IN2O3 and molecular weight 370.14 g/mol.
  • Suitable for linker attachment and synthesis of bifunctional degraders.