Learn More
Medchemexpress LLC 2,6-piperidinedione, 3-(1,3-dihydro-5-iodo-1-oxo-2H-isoindol-2-yl)- | 2291364-01-5 | MFCD32853043 | 98.0% | 370.14 g/mol | C13H11IN2O3 | 500 MG

Supplier: Medchemexpress LLC HYW441271500MG
Deoxy-thalidomide-5-iodine is a thalidomide analog used as a cereblon-binding ligand scaffold in PROTAC and chemical biology research. Provided as a research-grade small molecule with reported purity suitable for biochemical and cell-based studies.
- Purity reported as 98.0%.
- Suitable for PROTAC ligand development and E3 ligase recruitment studies.
- Iodinated scaffold enables further functionalization or conjugation chemistry.
- Available in multiple pack sizes, including 500 mg, for scale flexibility.
- Molecular formula C13H11IN2O3; molecular weight 370.14 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.