Learn More
Medchemexpress LLC 2,6-Piperidinedione, 3-(4-bromo-2,3-dihydro-3-methyl-2-oxo-1H-benzimidazol-1-yl)- | 2304754-51-4 | 99.8% | C13H12BrN3O3 | 250 MG
SDP
Catalog No. 5000358594
Encompass_Preferred
E3 ligase Ligand 63 is an E3 ligase ligand designed for the synthesis of CDK2 degrader 2. It functions within the ubiquitin-proteasome system, a cellular pathway exploited by PROTACs to selectively degrade target proteins.
  • Designed for synthesizing CDK2 degrader 2
  • Functions as an E3 ligase ligand
  • Exploits the ubiquitin-proteasome system for protein degradation
  • High purity of 99.8%
  • Stable for extended periods under recommended storage conditions
Catalog No. 50-003-58594 Supplier Medchemexpress LLC Supplier No. 250MGHY45512
E3 ligase Ligand 63 is an E3 ligase ligand designed for the synthesis of CDK2 degrader 2. It functions within the ubiquitin-proteasome system, a cellular pathway exploited by PROTACs to selectively degrade target proteins.
  • Designed for synthesizing CDK2 degrader 2
  • Functions as an E3 ligase ligand
  • Exploits the ubiquitin-proteasome system for protein degradation
  • High purity of 99.8%
  • Stable for extended periods under recommended storage conditions