Learn More
Medchemexpress LLC 2,6-piperidinedione, 3-(4-bromo-7-fluoro-1,3-dihydro-1-oxo-2H-isoindol-2-yl) | 2438239-34-8 | MFCD34576717 | 97.0% | 341.13 | C13H10BrFN2O3 | 250 MG
SDP
Catalog No. 5000284000
Encompass_Preferred
Lenalidomide-4-Br-7-fluoro is a synthetic derivative of lenalidomide bearing bromine and fluorine substituents on the benzyl ring. It is used as an E3 ligase ligand scaffold in PROTAC development and related chemical biology research, providing a modified cereblon-binding core for targeted protein degradation studies.
  • Derivative of lenalidomide with bromine and fluorine substituents on the benzyl ring.
  • Used as an E3 ligase ligand scaffold for PROTAC and chemical biology research.
  • Chemical formula C13H10BrFN2O3 and molecular weight 341.13 Da.
  • Identified by CAS number 2438239-34-8 and characterized by analytical data.
  • Typical batch NMR purity ~97.0% (example batch).
  • Store powder at -20°C for long-term stability; follow solvent storage recommendations for solutions.
Catalog No. 50-002-84000 Supplier Medchemexpress LLC Supplier No. HYW460227250MG
Lenalidomide-4-Br-7-fluoro is a synthetic derivative of lenalidomide bearing bromine and fluorine substituents on the benzyl ring. It is used as an E3 ligase ligand scaffold in PROTAC development and related chemical biology research, providing a modified cereblon-binding core for targeted protein degradation studies.
  • Derivative of lenalidomide with bromine and fluorine substituents on the benzyl ring.
  • Used as an E3 ligase ligand scaffold for PROTAC and chemical biology research.
  • Chemical formula C13H10BrFN2O3 and molecular weight 341.13 Da.
  • Identified by CAS number 2438239-34-8 and characterized by analytical data.
  • Typical batch NMR purity ~97.0% (example batch).
  • Store powder at -20°C for long-term stability; follow solvent storage recommendations for solutions.