Learn More
Medchemexpress LLC 2,6-Pyridinedicarboxamide, 4-(2-aminoethoxy)-N2,N6-bis[4-(2-aminoethoxy)-2-quinolinyl]-, hydrochloride (1:5) | 1781882-65-2 | C31H37Cl5N8O5 | 50 MG
SDP
Catalog No. 5000374849
Encompass_Preferred
Pyridostatin hydrochloride is a G-quadruplex DNA stabilizing agent that targets DNA and RNA G4s in cells. It promotes growth arrest in human cancer cells by inducing replication- and transcription-dependent DNA damage. This compound targets the proto-oncogene Src, reducing SRC protein levels and SRC-dependent cellular motility in human breast cancer cells. It is suitable for research use only.
  • Selective G-quadruplex DNA-binding small molecule
  • Induces cell cycle arrest in over 60 different cancer cell lines
  • Causes neurite retraction and synaptic loss
  • Downregulates BRCA1 protein at the transcriptional level
Catalog No. 50-003-74849 Supplier Medchemexpress LLC Supplier No. HY15176A50MG
Pyridostatin hydrochloride is a G-quadruplex DNA stabilizing agent that targets DNA and RNA G4s in cells. It promotes growth arrest in human cancer cells by inducing replication- and transcription-dependent DNA damage. This compound targets the proto-oncogene Src, reducing SRC protein levels and SRC-dependent cellular motility in human breast cancer cells. It is suitable for research use only.
  • Selective G-quadruplex DNA-binding small molecule
  • Induces cell cycle arrest in over 60 different cancer cell lines
  • Causes neurite retraction and synaptic loss
  • Downregulates BRCA1 protein at the transcriptional level