Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,6-Pyridinedicarboxylic acid

Catalog No. P6380825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P6380825G 25 g
P63808100G 100 g
P63808500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P6380825G Supplier Sigma Aldrich Supplier No. P6380825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,6-Pyridinedicarboxylic acid

Specifications

CAS 499-83-2
Melting Point 248°C to 250°C (dec.) (lit.)
Molecular Formula C7H5NO4
Linear Formula C7H5NO4
MDL Number MFCD00006299
Quantity 25 g
Synonym DPA; DPAc; Dipicolinic acid
Molecular Weight (g/mol) 167.12
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.