Learn More
2,6-Pyridinedicarboxylic acid, ≥99.5% (T), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Melting Point | 248°C to 250°C |
| CAS | 499-83-2 |
| Flash Point | 188°C - Closed Cup |
| Linear Formula | C7H5 NO4 |
| Quantity | 5 g |
| Solubility Information | Water: 1%, clear |
| Molecular Weight (g/mol) | 167.12 |
| Formula Weight | 167.12 |
| Percent Purity | ≥99.5% (T) |
| Grade | For ion chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.