Learn More
Optimize Technologies EXP™2 Replacement Stems, 2.6µL, 2.7µm HALO, Peptide ES-C18, 3pk
SDP
Supplier:  Optimize Technologies 1504020HN
Encompass_Preferred
Specialized features, patented technology, precision engineering and state-of-the-art manufacturing make the new EXP2 Stem Trap an unbeatable choice for ultra high-pressure trapping applications. Its slim architecture allows it to easily fit into crowded instrument compartments or to connect directly to tightly-spaced injection ports. When tightened by hand, the EXP2 Stem Trap seals to 8,700+ psi. Additionally, the product can be wrench tightened to 20,000+ psi (1,400+ bar). The unique packed floating stem installs directly into any 10-32 port and automatically adjusts to provide a perfect ZDV connection. The new EXP2 design incorporates an integral ferrule into the front-end of the holder so there is no separate ferrule to lose! The replacement stems are also low volume/ low-dispersion, intended for many repeat uses and is available with various bed volume and custom packing options. This product is recommended for nano flow applications. For use with EXP™2 Stem Holder Only
Catalog No. 50-200-6472