Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-([7-(acetylamino)-6-(benzyloxy)-2-phenylperhydropyrano[3,2-d][1,3]dioxin-8-yl]oxy)propanoic acid, Thermo Scientific™

Catalog No. RH01183DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RH01183DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RH01183DA Supplier Thermo Scientific Chemicals Supplier No. RH01183DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 499104-69-7
Molecular Formula C25H29NO8
Molecular Weight (g/mol) 471.506
InChI Key JPPMVSNCFXDOJX-UHFFFAOYSA-N
Synonym 2-7-acetylamino-6-benzyloxy-2-phenylperhydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2r-2-4ar,6s,7r,8r,8as-7-acetamido-6-benzyloxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2-6-benzyloxy-7-acetamido-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, benzyl n-acetyl-4,6-o-benzylidenemuramic acid, 2-7-acetamido-6-benzyloxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2-7-acetamido-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid
PubChem CID 3843768
IUPAC Name 2-[(7-acetamido-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-8-yl)oxy]propanoic acid
SMILES CC(C(=O)O)OC1C(C(OC2C1OC(OC2)C3=CC=CC=C3)OCC4=CC=CC=C4)NC(=O)C

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 471.5
Percent Purity 97%
Chemical Name or Material 2-{[7-(Acetylamino)-6-(benzyloxy)-2-phenylperhydropyrano[3, 2-d][1, 3]dioxin-8-yl]oxy}propanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.