Learn More
Medchemexpress LLC 2,7-bis(3-morpholinopropyl)-4-((2-(pyrrolidin-1-yl)ethyl)amino)benzo[lmn][3,8]phenanthroline | 2101208-44-8 | 98.2% | 50 MG
SDP
Catalog No. 5000230289
Encompass_Preferred
CM03 is a small-molecule G-quadruplex (G4) DNA ligand that binds and stabilizes G4 structures, downregulates G4-containing genes, and increases double-strand break events. It has demonstrated selective potency in pancreatic cancer cell models and is supplied for research use only.
  • Binds and stabilizes DNA G-quadruplex structures.
  • Downregulates expression of G4-containing genes.
  • Increases incidence of double-strand DNA breaks due to torsional strain.
  • Molecular formula C34H44N6O6; molecular weight 632.75 g/mol.
  • High purity suitable for research applications.
  • Intended for research use only; not for human use.
Catalog No. 50-002-30289 Supplier Medchemexpress LLC Supplier No. HY12186250MG
CM03 is a small-molecule G-quadruplex (G4) DNA ligand that binds and stabilizes G4 structures, downregulates G4-containing genes, and increases double-strand break events. It has demonstrated selective potency in pancreatic cancer cell models and is supplied for research use only.
  • Binds and stabilizes DNA G-quadruplex structures.
  • Downregulates expression of G4-containing genes.
  • Increases incidence of double-strand DNA breaks due to torsional strain.
  • Molecular formula C34H44N6O6; molecular weight 632.75 g/mol.
  • High purity suitable for research applications.
  • Intended for research use only; not for human use.