Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-dihexylfluorene 98.0+%, TCI America™
SDP

Catalog No. B40291G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B40291G Supplier TCI America Supplier No. B40291G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 254755-24-3
Molecular Formula C37H56B2O4
Molecular Weight (g/mol) 586.471
MDL Number MFCD16621093
InChI Key SYMMYBWUPCWTEI-UHFFFAOYSA-N
Synonym 9,9-Dihexylfluorene-2,7-diboronic Acid Bis(pinacol) Ester
PubChem CID 20582027
IUPAC Name 2-[9,9-dihexyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(C3(CCCCCC)CCCCCC)C=C(C=C4)B5OC(C(O5)(C)C)(C)C

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 1 g
Formula Weight 586.47
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-dihexylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.