Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromo-9,9-bis(6-bromohexyl)fluorene 98.0+%, TCI America™
SDP

Catalog No. B35525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35525G 5 g
B35521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35525G Supplier TCI America Supplier No. B35525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 570414-33-4
Molecular Formula C25H30Br4
Molecular Weight (g/mol) 650.131
MDL Number MFCD09260450
InChI Key OJMAUBALNSWGDC-UHFFFAOYSA-N
PubChem CID 21867930
IUPAC Name 2,7-dibromo-9,9-bis(6-bromohexyl)fluorene
SMILES C1=CC2=C(C=C1Br)C(C3=C2C=CC(=C3)Br)(CCCCCCBr)CCCCCCBr

Specifications

Melting Point 72°C
Color White
Quantity 5 g
Formula Weight 650.13
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9,9-bis(6-bromohexyl)fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.