Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromo-9,9'-spirobi[9H-fluorene] 98.0+%, TCI America™
SDP

Catalog No. D37811G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D37811G Supplier TCI America Supplier No. D37811G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 171408-84-7
Molecular Formula C25H14Br2
Molecular Weight (g/mol) 474.20
MDL Number MFCD08704218
InChI Key UPJLZKCEPFAKSH-UHFFFAOYSA-N
PubChem CID 15544767
IUPAC Name 2,7-dibromo-9,9'-spirobi[fluorene]
SMILES BrC1=CC2=C(C=C1)C1=C(C=C(Br)C=C1)C21C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 474.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9,9′-spirobi[9H-fluorene]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.