Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromo-9-hexylcarbazole 98.0+%, TCI America™
SDP

Catalog No. D50555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50555G 5 g
D50551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50555G Supplier TCI America Supplier No. D50555G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 654676-12-7
Molecular Formula C18H19Br2N
Molecular Weight (g/mol) 409.165
InChI Key YSUUFISKPJBPOH-UHFFFAOYSA-N
PubChem CID 25150458
IUPAC Name 2,7-dibromo-9-hexylcarbazole
SMILES CCCCCCN1C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 5 g
Formula Weight 409.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9-hexylcarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.