Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,7-dichloro-9,9-dimethyl-10-[2-(2H-tetrazol-5-yl)ethyl]acridine | 1443290-89-8 | 99.3% | 1 ML
SDP

Catalog No. 5000233189
Encompass_Preferred

ML67-33 (IUPAC: 2,7-dichloro-9,9-dimethyl-10-[2-(2H-tetrazol-5-yl)ethyl]acridine) is a small-molecule activator of temperature- and mechano-sensitive two-pore domain (K2P) potassium channels used as a research tool to modulate TREK channel activity in neurophysiology and ion channel studies.

  • Selective activator of K2P/TREK channels.
  • High purity (99.3% by HPLC).
  • Molecular weight 374.27 g/mol; formula C18H17Cl2N5.
  • Supplied as 10 mM solution in DMSO, 1 ML.
  • Storage: powder at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).
  • For research use only; not intended for human or clinical use.

Catalog No. 50-002-33189 Supplier Medchemexpress LLC Supplier No. HY12034810MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ML67-33 (IUPAC: 2,7-dichloro-9,9-dimethyl-10-[2-(2H-tetrazol-5-yl)ethyl]acridine) is a small-molecule activator of temperature- and mechano-sensitive two-pore domain (K2P) potassium channels used as a research tool to modulate TREK channel activity in neurophysiology and ion channel studies.

  • Selective activator of K2P/TREK channels.
  • High purity (99.3% by HPLC).
  • Molecular weight 374.27 g/mol; formula C18H17Cl2N5.
  • Supplied as 10 mM solution in DMSO, 1 ML.
  • Storage: powder at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).
  • For research use only; not intended for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.