Learn More
Cayman Chemical 2,7-Dichlorodihydrofluorescein diacetate, 4091-99-0, 100 mg
SDP
Supplier:  Cayman Chemical 85155100
Encompass_Preferred
A fluorescent indicator of peroxynitrite formation; neither NO, superoxide, nor hydrogen peroxide alone appear to oxidize DCFH
Catalog No. NC1476516