Learn More
Cayman Chemical 2,7-Dichlorodihydrofluorescein diacetate, 4091-99-0, 50 mg
SDP
Supplier:  Cayman Chemical 8515550
Encompass_Preferred
Molecular Formula C24H16Cl2O7, Purity ≥95%, Formula Weight 487.3, Synonyms: 2-(2,7-dichloro-3,6-diacetyloxy-9H-xanthen-9-yl)-benzoic acid, DCFH, DCFH-DA; A fluorescent indicator of peroxynitrite formation; neither NO, superoxide, nor hydrogen peroxide alone appear to oxidize DCFH.
Catalog No. NC0227997