Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2',7'-Dichlorodihydrofluorescein diacetate | 4091-99-0 | 99.8% | 250 MG
SDP

Catalog No. 5000413737
Encompass_Preferred

H2DCFDA is a cell-permeable probe utilized for detecting intracellular reactive oxygen species (ROS) with an excitation/emission (Ex/Em) of 488/525 nm. It targets Reactive Oxygen Species (ROS) and acts as a Fluorescent Dye, involved in Immunology/Inflammation, Metabolic Enzyme/Protease, and NF-κB pathways.

  • Cell-permeable probe for detecting intracellular reactive oxygen species.
  • Features an excitation/emission of 488/525 nm.
  • Molecular formula is C24H16Cl2O7.
  • Molecular weight is 487.29.
  • Soluble in DMSO at 250 mg/mL.
  • Recommended protocols for solution preparation and cell staining.

Catalog No. 50-004-13737 Supplier Medchemexpress LLC Supplier No. 250MGHYD0940
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H2DCFDA is a cell-permeable probe utilized for detecting intracellular reactive oxygen species (ROS) with an excitation/emission (Ex/Em) of 488/525 nm. It targets Reactive Oxygen Species (ROS) and acts as a Fluorescent Dye, involved in Immunology/Inflammation, Metabolic Enzyme/Protease, and NF-κB pathways.

  • Cell-permeable probe for detecting intracellular reactive oxygen species.
  • Features an excitation/emission of 488/525 nm.
  • Molecular formula is C24H16Cl2O7.
  • Molecular weight is 487.29.
  • Soluble in DMSO at 250 mg/mL.
  • Recommended protocols for solution preparation and cell staining.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.