Learn More
TARGETMOL CHEMICALS INC Research Chemical 500mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T20684500MG
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg, 200 mg, 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 2',7'-Dichlorofluorescein diacetate (Diacetyldichlorofluorescein), a cell-permeable fluorogenic probe, is useful for the detection of nitric oxide (NO) and reactive oxygen species (ROS) and for the determination of the degree of overall oxidative stress. purity: 99%
Catalog No. 50-303-8145