Learn More
Abcam 2', 7'-Dichlorofluorescein di-ß-D-lactoside, 5MG
Catalog No. 502671852
Encompass_Preferred
MW 1049.76 g/mol. Highly sensitive fluorescent substrate for measuring beta-lactosidase and glucosidase inside live cells. Retains fluorescence at low pH due to lower pKa than comparable fluorophores. Suitable for measuring activity in the acidic lysosome environment.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1852 Supplier Abcam Supplier No. AB2753665MG
MW 1049.76 g/mol. Highly sensitive fluorescent substrate for measuring beta-lactosidase and glucosidase inside live cells. Retains fluorescence at low pH due to lower pKa than comparable fluorophores. Suitable for measuring activity in the acidic lysosome environment.
The product is subject to the following: Abcam Restricted Use Statement