Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',7'-Dichlorofluorescein, Sigma-Aldrich

Catalog No. 01258847 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-847 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-847 Supplier MilliporeSigma Supelco Supplier No. 41021710G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2',7'-Dichlorofluorescein is an oxidation-sensitive fluorescent probe for reactive oxygen species studies.

  • Used to investigate reactive oxygen species formation in cells.
  • Can serve as a fluorogenic substrate in peroxidase-activity studies.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-54-0

Specifications

Formulation Powder
Quantity 10 g
Product Type Stain
Molecular Weight (g/mol) 401.2 g/mol
Label Type Other Labels or Dyes
Color Red-Orange
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.