Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,7-Difluoro-9-Mesityl-10-Methylacridinium Tetrafluoroborate

Catalog No. 909254250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-925-4250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-925-4250MG Supplier Sigma Aldrich Supplier No. 909254250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Robust acridinium-based photocatalyst for use as an alternative to transition-metal-based photocatalysts with higher chemical stability and attenuated redox potential.

Specifications

CAS 69075-42-9
Linear Formula C11H12N2O6
Quantity 50 mg
Synonym 17-Azido-N-(2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)-3,6,9,12,15-pentaoxaheptadecanamide; Crosslinker-E3 Ligase ligand conjugate; Pomalidomide conjugate; Protein degrader building block for PROTAC(R) research; Template for synthesis of targeted protein degrader
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.