Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dihydroxy-9H-fluoren-9-one 98.0+%, TCI America™
SDP

Catalog No. D16855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16855G 5 g
D168525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16855G Supplier TCI America Supplier No. D16855G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42523-29-5
Molecular Formula C13H8O3
Molecular Weight (g/mol) 212.20
MDL Number MFCD00037141
InChI Key CWHPQXRTQSNTRR-UHFFFAOYSA-N
PubChem CID 4354181
IUPAC Name 2,7-dihydroxy-9H-fluoren-9-one
SMILES OC1=CC2=C(C=C1)C1=C(C=C(O)C=C1)C2=O

Specifications

Melting Point 329°C
Quantity 5 g
Formula Weight 212.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dihydroxy-9H-fluoren-9-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.