Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dihydroxynaphthalene, MP Biomedicals

Catalog No. ICN206022 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN206022 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN206022 Supplier MP Biomedicals, Inc Supplier No. 0520602205

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 582-17-2
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004085
InChI Key DFQICHCWIIJABH-UHFFFAOYSA-N
Synonym 2,7-dihydroxynaphthalene, 2,7-naphthalenediol, naphthalenediol-2,7, unii-0to8e448ud, naphthalenediol-2,7 french, 2,7-dihydroxy naphthalene, 2,7-naphthalene diol, naphthalene-2-7-diol, 2,7-dihdroxynaphthalene
PubChem CID 11397
IUPAC Name naphthalene-2,7-diol
SMILES OC1=CC2=CC(O)=CC=C2C=C1

Specifications

Quantity 5 g
Formula Weight 160.17
Chemical Name or Material 2, 7-Dihydroxynaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.